Learn More
Grainger Fuse Block 0 to 30A 240VAC
Catalog No. 502752666
Encompass_Preferred
The BUSSMANN Finger-Safe Fuse Block with Cover is a 30A, 600VAC finger-safe fuse block that provides overcurrent protection. It has screw-type terminals that accept up to #14 to #6 AWG wire. The clear cover allows for fuse inspection while preventing accidental contact. This UL listed block can be mounted to panels or walls with the included mounting hardware. With dimensions of 2-1/2" length x 2-1/8" width x 2-3/8" height, this fuse block provides safe and convenient fuse access.
Catalog No. 50-275-2666 Supplier Grainger Supplier No. 14L603
The BUSSMANN Finger-Safe Fuse Block with Cover is a 30A, 600VAC finger-safe fuse block that provides overcurrent protection. It has screw-type terminals that accept up to #14 to #6 AWG wire. The clear cover allows for fuse inspection while preventing accidental contact. This UL listed block can be mounted to panels or walls with the included mounting hardware. With dimensions of 2-1/2" length x 2-1/8" width x 2-3/8" height, this fuse block provides safe and convenient fuse access.