Learn More
Grainger Fuse Block 0 to 30A 600VAC
Catalog No. 502753287
Encompass_Preferred
The Bussmann Finger-Safe Fuse Block with Cover is a 30A, 600VAC finger-safe fuse block that provides overcurrent protection. It has screw terminals that accept up to #14 to #6 AWG wire and features a clear cover for safety. This UL listed block can be mounted to panels or walls and is made of durable thermoplastic. The maximum fuse size it accepts is 30A.
Catalog No. 50-275-3287 Supplier Grainger Supplier No. 14L601
The Bussmann Finger-Safe Fuse Block with Cover is a 30A, 600VAC finger-safe fuse block that provides overcurrent protection. It has screw terminals that accept up to #14 to #6 AWG wire and features a clear cover for safety. This UL listed block can be mounted to panels or walls and is made of durable thermoplastic. The maximum fuse size it accepts is 30A.