Learn More
Grainger Fuse Block 0 to 30A 600VAC
Catalog No. 502748684
Encompass_Preferred
The BUSSMANN Fuse Block is an industrial fuse block that fits up to 6 fuses and has a maximum voltage of 600VAC. It has screw terminals that accept up to #14 to #10 AWG wire. The block measures 2-1/4"W x 2-3/4"H x 2-3/4"D and is made of thermoplastic. This fuse block can be mounted to walls or panels using the included mounting hardware. It is UL listed and CSA certified. The BUSSMANN Fuse Block provides overload protection for industrial control panels and machinery.
Catalog No. 50-274-8684 Supplier Grainger Supplier No. 14L609
The BUSSMANN Fuse Block is an industrial fuse block that fits up to 6 fuses and has a maximum voltage of 600VAC. It has screw terminals that accept up to #14 to #10 AWG wire. The block measures 2-1/4"W x 2-3/4"H x 2-3/4"D and is made of thermoplastic. This fuse block can be mounted to walls or panels using the included mounting hardware. It is UL listed and CSA certified. The BUSSMANN Fuse Block provides overload protection for industrial control panels and machinery.