Learn More
Grainger Fuse Block 31 to 60A 600VAC
Catalog No. 502748709
Encompass_Preferred
The Bussmann Finger-Safe Fuse Block with Cover is a fuse block that provides overcurrent protection for electrical circuits. It has space for up to 6 ATC/ATO automotive blade fuses and features a clear plastic hinged cover for safety. The maximum voltage is 32V DC and maximum current is 60A. The fuse block has tin-plated brass terminals and measures 2-3/4" L x 1-3/4" W x 1-3/4" H. This finger-safe fuse block is UL recognized and CSA certified. The cover helps prevent accidental contact with live parts.
Catalog No. 50-274-8709 Supplier Grainger Supplier No. 14L613
The Bussmann Finger-Safe Fuse Block with Cover is a fuse block that provides overcurrent protection for electrical circuits. It has space for up to 6 ATC/ATO automotive blade fuses and features a clear plastic hinged cover for safety. The maximum voltage is 32V DC and maximum current is 60A. The fuse block has tin-plated brass terminals and measures 2-3/4" L x 1-3/4" W x 1-3/4" H. This finger-safe fuse block is UL recognized and CSA certified. The cover helps prevent accidental contact with live parts.