Learn More
Grainger Fuse Block 61 to 100A 600VAC
Catalog No. 502749702
Encompass_Preferred
The Bussmann Fuse Block is an industrial fuse block that fits up to 6 fuses and has a maximum voltage of 600VAC. It has screw terminals that accept wire sizes from #14-#10 AWG. The block measures 2-1/4"W x 2-3/4"H x 2-3/4"D and is made of thermoplastic. This UL listed block can be mounted to walls or panels using the included mounting hardware. With its high short circuit rating, it provides overcurrent protection for industrial control panels and machinery.
Catalog No. 50-274-9702 Supplier Grainger Supplier No. 14L614
The Bussmann Fuse Block is an industrial fuse block that fits up to 6 fuses and has a maximum voltage of 600VAC. It has screw terminals that accept wire sizes from #14-#10 AWG. The block measures 2-1/4"W x 2-3/4"H x 2-3/4"D and is made of thermoplastic. This UL listed block can be mounted to walls or panels using the included mounting hardware. With its high short circuit rating, it provides overcurrent protection for industrial control panels and machinery.