Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Ceramic Fuse 10A, 250V

Catalog No. NC9456276 Shop All Thermo Scientific Products
Encompass_Preferred

Ceramic Fuse 10A, 250V.

Catalog No. NC9456276 Supplier Thermo Scientific Supplier No. 230066

missing translation for 'validMainframeMsg'

Ceramic Fuse 10A, 250V.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.