Learn More
MSC Ideal 34-003 11-3/4 Inch Long, Glass Filled Polypropylene, Insulated Fuse Puller For Use with 0 to 100 Amp, 250 Volt Fuses, 1/2 to 1 Inch Diameter Fuses, 600 Volt Fuses, Cartridge Fuses
Catalog No. 501305076
Encompass
Ideal 34-003 11-3/4 Inch Long, Glass Filled Polypropylene, Insulated Fuse Puller For Use with 0 to 100 Amp, 250 Volt Fuses, 1/2 to 1 Inch Diameter Fuses, 600 Volt Fuses, Cartridge Fuses
Catalog No. 50-130-5076 Supplier MSC Supplier No. 54043088
Ideal 34-003 11-3/4 Inch Long, Glass Filled Polypropylene, Insulated Fuse Puller For Use with 0 to 100 Amp, 250 Volt Fuses, 1/2 to 1 Inch Diameter Fuses, 600 Volt Fuses, Cartridge Fuses