Learn More
Elemental Scientific Inc FUSE SILICA OBST REMVL KIT
SDP
Catalog No. NC0632272
Encompass_Preferred
Fused Silica Obstruction Removal Kit for ST type Nebulizers. Includes one guide, 10 pieces of fused silica and instructions.
Catalog No. NC0632272 Supplier Elemental Scientific Inc Supplier No. ES20460001
Fused Silica Obstruction Removal Kit for ST type Nebulizers. Includes one guide, 10 pieces of fused silica and instructions.