Learn More
Waters Corp Waters Tubing, Fused Silica, ApCI Probe
Catalog No. 50466548 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Tubing, Fused Silica, ApCI Probe is a spare part kit for mass spectrometry applications, specifically for Atmospheric Pressure Chemical Ionization (APCI) probes. This tubing is made of fused silica, a high-purity glass material known for its excellent optical properties and chemical inertness, making it suitable for sensitive analytical procedures.
  • Ensures high performance in mass spectrometry applications.
  • Constructed from fused silica for chemical inertness and durability.
  • Designed for use with ApCI probes.
  • Ideal for maintaining and repairing analytical equipment.
Catalog No. 50-466-548 Supplier Waters Corp Supplier No. 430000109
Waters Tubing, Fused Silica, ApCI Probe is a spare part kit for mass spectrometry applications, specifically for Atmospheric Pressure Chemical Ionization (APCI) probes. This tubing is made of fused silica, a high-purity glass material known for its excellent optical properties and chemical inertness, making it suitable for sensitive analytical procedures.
  • Ensures high performance in mass spectrometry applications.
  • Constructed from fused silica for chemical inertness and durability.
  • Designed for use with ApCI probes.
  • Ideal for maintaining and repairing analytical equipment.