Learn More
Thermo Scientific Fused Silica Optical Fiber 100 Feet
Supplier:  Thermo Scientific 0010610498
Encompass
Fused silica tubing with polyimide outer coating provides inert, high temperature-resistant sample transfer line between atmospheric pressure chemical ionization source and mass analyzer for Fourier transform ion cyclotron resonance mass spectrometry, enabling sensitive, soft ionization analysis of thermally labile and low polarity compounds.
Catalog No. NC9549136