Learn More
Perkin Elmer US LLC Fused Silica Obstruction Removal Kit for ST Type Nebulizers
Catalog No. 502105100
Encompass_Preferred
Includes one guide, 10 pieces of fused silica and instructions. Note: Never back-flush a PFA-ST nebulizer.
Catalog No. 50-210-5100 Supplier Perkin Elmer US LLC Supplier No. N8145236
Includes one guide, 10 pieces of fused silica and instructions. Note: Never back-flush a PFA-ST nebulizer.