Learn More
Elemental Scientific Inc FUSED SILICA REFILL KIT
SDP
Catalog No. NC0353569
Encompass_Preferred
120 um o.d. fused silica refill for Obstruction Removal Kit For use with ST type Nebulizers only. 10 pieces
Catalog No. NC0353569 Supplier Elemental Scientific Inc Supplier No. ES20460002
120 um o.d. fused silica refill for Obstruction Removal Kit For use with ST type Nebulizers only. 10 pieces