Learn More
Waters Corp Tubing, Fused Silica, 25 µm ID, 1 m
Catalog No. 50792153 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Fused Silica Tube, 25µm ID is used for the Stream Select Module in the CapLC XE System. It is also used in the SYNAPT G2 HDMS and SYNAPT G2 MS. The capillary measures 375µm x25µm and is 1 m in length.
Catalog No. 50-792-153 Supplier Waters Corp Supplier No. 6028622S1
The Fused Silica Tube, 25µm ID is used for the Stream Select Module in the CapLC XE System. It is also used in the SYNAPT G2 HDMS and SYNAPT G2 MS. The capillary measures 375µm x25µm and is 1 m in length.