Learn More
Waters Corp Waters Corporation Tubing, Fused Silica, 375 μm x 75 μm, 1 m
Catalog No. 50792299 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This fused silica tubing is designed for various laboratory applications, offering precise dimensions for reliable performance. It is a critical spare part for the NanoLockspray Option, ensuring continued functionality of the system.
  • Made from fused silica for high performance
  • Measures 375 μm x 75 μm x 1 m
  • Serves as a spare part for the NanoLockspray Option
  • Comes as 1 unit per package
  • Replacement is recommended as required
Catalog No. 50-792-299 Supplier Waters Corp Supplier No. 6490512S1
This fused silica tubing is designed for various laboratory applications, offering precise dimensions for reliable performance. It is a critical spare part for the NanoLockspray Option, ensuring continued functionality of the system.
  • Made from fused silica for high performance
  • Measures 375 μm x 75 μm x 1 m
  • Serves as a spare part for the NanoLockspray Option
  • Comes as 1 unit per package
  • Replacement is recommended as required