Learn More
Analytical Sales & Services Fused Silica Tubing, 360µm x 100um x 2M
Catalog No. 5000807083
Encompass
Fused Silica Tubing - 360µm (0.0145") OD x 100µm ID x 2M Length Pressure Rating: 10,000psi (690 bar) Color: Natural
Catalog No. 50-008-07083 Supplier Analytical Sales & Services Supplier No. 1082
Add to Cart
Add to Cart
Fused Silica Tubing - 360µm (0.0145") OD x 100µm ID x 2M Length Pressure Rating: 10,000psi (690 bar) Color: Natural