Learn More
Analytical Sales & Services Fused Silica Tubing, 360µm x 20µm x 2M
Catalog No. 5000806033
Encompass
Fused Silica Tubing - 360µm (0.0145") OD x 20µm ID x 2M Length Pressure Rating: 10,000psi (690 bar) Color: Natural
Catalog No. 50-008-06033 Supplier Analytical Sales & Services Supplier No. 1052
Fused Silica Tubing - 360µm (0.0145") OD x 20µm ID x 2M Length Pressure Rating: 10,000psi (690 bar) Color: Natural