Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fusidic Acid Sodium, G-Biosciences
SCI_ED

Catalog No. S162514
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162514 1 g
S000001 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S162514 Supplier G-Biosciences Supplier No. RC1030
Add to Cart
Add to Cart

Fusidic acid sodium is a bacteriostatic steroid antibiotic targeting gram-positive bacteria, especially those responsible for skin infections.

It has been shown to inhibit nitric oxide lysis of islet cells, cytokine production, neutrophils, and T cells. It is highly water soluble. Fusidic acid sodium inhibits protein synthesis in prokaryotes via suppression of elongation factor G.

Chemical Identifiers

Molecular Weight (g/mol) 538.69 g/mol

Specifications

Boiling Point 521.49°C
Quantity 1 g
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.