Learn More
Enzo Life Sciences Fusidic acid. sodium salt (50mg). CAS: 751-94-0
SDP
Catalog No. 502008906
Encompass_Preferred
Antibiotic which suppresses nitric oxide (NO) toxicity in pancreatic islet cells. Inhibits protein synthesis by inhibition of elongation factor G at the level of the ribosome. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8906 Supplier Enzo Life Sciences Supplier No. ALX380011M050
Add to Cart
Add to Cart
Antibiotic which suppresses nitric oxide (NO) toxicity in pancreatic islet cells. Inhibits protein synthesis by inhibition of elongation factor G at the level of the ribosome. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.