Learn More
Medchemexpress LLC Fusidic acid sodium salt | 751-94-0 | 99.6% | 5 G
SDP
Catalog No. 5000413684
Encompass_Preferred
Fusidic acid sodium salt is an orally available antibacterial agent that inhibits bacterial protein synthesis. It also exhibits antidiabetic effects in a rat model and improves colitis symptoms in rats.
  • Inhibits bacterial protein synthesis by preventing the release of translation elongation factor G (EF-G) from ribosomes.
  • Exhibits antidiabetic effects by inhibiting the effects of interleukins IL-1 and IL-6 on glucose-induced insulin production.
  • Improves colitis symptoms.
  • Inhibits the growth of *Toxoplasma gondii* and *Listeria monocytogenes* EGD in vitro.
Catalog No. 50-004-13684 Supplier Medchemexpress LLC Supplier No. HYB1350A5G
Fusidic acid sodium salt is an orally available antibacterial agent that inhibits bacterial protein synthesis. It also exhibits antidiabetic effects in a rat model and improves colitis symptoms in rats.
  • Inhibits bacterial protein synthesis by preventing the release of translation elongation factor G (EF-G) from ribosomes.
  • Exhibits antidiabetic effects by inhibiting the effects of interleukins IL-1 and IL-6 on glucose-induced insulin production.
  • Improves colitis symptoms.
  • Inhibits the growth of *Toxoplasma gondii* and *Listeria monocytogenes* EGD in vitro.