Learn More
Cayman Chemical Fusidic Acid (sodium salt), Purity: ≥98%, CAS: 751-94-0, 100 mg
SDP
Catalog No. 502969832
Encompass_Preferred
A fusidane antibiotic; active against a variety of Gram-positive bacteria (MIC50s = 0.01-20 µg/ml) but not Gram-negative bacteria or fungi (MIC50s = >100 µg/ml for all); inhibits EF-G-mediated ribosomal recycling and protein translocation in isolated E. coli ribosomes (IC50s = ~0.1 and ~200 µM, respectively); reduces the number of skin CFUs and levels of TNF-α and IL-6 in a mouse model of MRSA skin wound infection
Catalog No. 50-296-9832 Supplier Cayman Chemical Supplier No. 14825100 MG
A fusidane antibiotic; active against a variety of Gram-positive bacteria (MIC50s = 0.01-20 µg/ml) but not Gram-negative bacteria or fungi (MIC50s = >100 µg/ml for all); inhibits EF-G-mediated ribosomal recycling and protein translocation in isolated E. coli ribosomes (IC50s = ~0.1 and ~200 µM, respectively); reduces the number of skin CFUs and levels of TNF-α and IL-6 in a mouse model of MRSA skin wound infection