Learn More
SOMNI Scientific™ Fusion 2™
SDP
Catalog No. 04731224 Shop All Somni Scientific Products
Change view
Click to view available options
Animal:
Mouse, Neonatal Rat
Mouse, Rat
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Animal
04-731-224 Mouse, Rat
04-731-223 Mouse, Neonatal Rat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 04-731-224 Supplier SOMNI Scientific™ Supplier No. SF22002
Add to Cart
Add to Cart
A new standard in anesthesia delivery
  • Integrates advanced low-flow control and active waste gas scavenging in a single system
  • Delivers precise, stable, and reproducible anesthetic output
  • Dual Flowmeters support two-station operation
  • Reduces carrier gas consumption
  • Single compact platform
  • Designed to improve efficiency
  • Reduced lab footprint
  • Enhanced user safety
  • Reduces waste anesthetic exposure
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Voltage 120 V
Animal Mouse, Rat
Type Anesthesia Machine