Learn More
Chemyx Inc Fusion 200-X Syringe Pump, 0.5µL to 60mL, 9.5 x 8.2 x 4.9 inches
Supplier:  Chemyx Inc 0720X
Encompass
The Fusion 200-X is a programmable, dual-channel infusion and withdrawal modular syringe pump. This advanced precision syringe pump features two syringes to infuse/withdraw simultaneously at pulseless, reproducible rates; a programmable multi-step mode that allows users to program complex multi-step flow rates; computer control with Lab view and other programs; an add-on 4-syringe push and pull rack for four large syringes; an add-on 11-syringe push and pull rack to increase the syringe number up to eleven. Fusion 200-X the full-glass touch screen is chemical resistant and easy to clean for lab environments of all types. This syringe pump is designed for precision dispensing/dilution, organ/tissue perfusion, and microfluidics, electrospinning, aerosols, pharmaceutical, and bioscience research.
Catalog No. NC2235109