Learn More
Haws™ Chrome-Plated brass Shower Ball Valve
SDP
Catalog No. 18888314 Shop All Haws Corporation Products
Change view
Click to view available options
Material:
Chrome-plated Brass, Stainless Steel Mesh
Catalog No. Material
18-888-314 Chrome-plated Brass, Stainless Steel Mesh
1 options
Catalog No. 18-888-314 Supplier Haws™ Supplier No. SP260.158
  • $640.00
    $508.65 / Each of 1
    Save $131.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Haws™ Chrome-Plated brass Shower Ball Valve is a stay-open ball valve, equipped with a stainless steel stem.
Model SP260.158 chrome-plated brass stay-open ball valve, equipped with a stainless steel stem. Full flow occurs with pull on lever. Special self-draining feature allows valve and connecting lines to showerhead to drain to prevent freezing. Full flow occurs with pull on lever. 1 in. NPT.
Warranty and Services
1 year from ship date (Limited)
Specifications
For Use With (Equipment) Emergency Shower and Eyewash
Material Chrome-plated Brass, Stainless Steel Mesh
Product Type Shower Ball Valve
Weight (English) 3 lb.
Weight (Metric) 1.36 kg
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov