Learn More
Aviva Systems Biology Corp SRSF10 Antibody - C-terminal region (ARP41082_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41082_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUSIP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human FUSIP1. Alternate names: FUSIP2, NSSR, SFRS13, SRp38, SRrp40, TASR, TASR1, TASR2, FUSIP1, SFRS13AApplicationIHC, WB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57714