Learn More
ATCC Enterocloster clostridioformis; Strain 0316 [T90]
Catalog No. 502380536
Encompass
Enterocloster clostridioformis strain VPI 0316 [T90] is an anaerobic bacterial type strain that was isolated from the rumen of a calf.
Catalog No. 50-238-0536 Supplier ATCC Supplier No. 25537
Enterocloster clostridioformis strain VPI 0316 [T90] is an anaerobic bacterial type strain that was isolated from the rumen of a calf.