Learn More
ATCC Fusobacterium nucleatum; Strain VPI 4355; genomic DNA
Catalog No. 502384218
Encompass
Genomic DNA from Fusobacterium nucleatum subsp. nucleatum strain VPI 4355 (ATCC 25586). This whole-genome sequenced product can be used in PCR or other molecular biology applications.
Catalog No. 50-238-4218 Supplier ATCC Supplier No. 25586D5
Genomic DNA from Fusobacterium nucleatum subsp. nucleatum strain VPI 4355 (ATCC 25586). This whole-genome sequenced product can be used in PCR or other molecular biology applications.