Learn More
ATCC Fusobacterium nucleatum; subsp. nucleatum; Strain VPI 4355
Supplier:  ATCC 25586
Encompass
Fusobacterium nucleatum subsp. nucleatum strain VPI 4355 [1612A] is a bacterial type strain that was isolated from a cervico-facial lesion. This strain is whole-genome sequenced and has applications in quality control, bioinformatics, and media testing.
Catalog No. 50-238-2575
 Restricted Item