Learn More
ATCC Bacillus thuringiensis
Supplier:  ATCC 33679
Encompass
Bacillus thuringiensis strain NRRL B-3792 is a whole-genome sequenced strain that was isolated from diseased insect larvae. This bacterial culture produces spores and toxic parasporal crystals.
Catalog No. 50-237-9694