Learn More
Medchemexpress LLC Fustin | 20725-03-5 | 1 MG
SDP
Catalog No. 5000417495
Encompass_Preferred
Fustin ((±)-Fustin; 3,7,3',4'-Tetrahydroxyflavanone) is a potent amyloid β (Aβ) inhibitor. It increases acetylcholine (ACh) levels, choline acetyltransferase (ChAT) activity, and ChAT gene induced by Aβ (1-42). It also decreases acetyl cholinesterase (AChE) activity and AChE gene expression induced by Aβ (1-42). Fustin increases muscarinic M1 receptor gene expression and muscarinic M1 receptor binding activity. It can be used for Alzheimer's disease research.
  • Potent amyloid β (Aβ) inhibitor.
  • Increases acetylcholine (ACh) levels and choline acetyltransferase (ChAT) activity.
  • Decreases acetylcholinesterase (AChE) activity.
  • Enhances muscarinic M1 receptor expression and binding.
  • Suitable for Alzheimer's disease research.
Catalog No. 50-004-17495 Supplier Medchemexpress LLC Supplier No. HYN83761MG
Fustin ((±)-Fustin; 3,7,3',4'-Tetrahydroxyflavanone) is a potent amyloid β (Aβ) inhibitor. It increases acetylcholine (ACh) levels, choline acetyltransferase (ChAT) activity, and ChAT gene induced by Aβ (1-42). It also decreases acetyl cholinesterase (AChE) activity and AChE gene expression induced by Aβ (1-42). Fustin increases muscarinic M1 receptor gene expression and muscarinic M1 receptor binding activity. It can be used for Alzheimer's disease research.
  • Potent amyloid β (Aβ) inhibitor.
  • Increases acetylcholine (ACh) levels and choline acetyltransferase (ChAT) activity.
  • Decreases acetylcholinesterase (AChE) activity.
  • Enhances muscarinic M1 receptor expression and binding.
  • Suitable for Alzheimer's disease research.