Learn More
Aviva Systems Biology Corp FUT1 Antibody - middle region (ARP44222_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44222_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FUT1. Alternate names: H, HH, HSCApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-130