Learn More
Aviva Systems Biology Corp FUT3 Antibody - C-terminal region (ARP63943_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63943_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUT3. It was validated on Western Blot. . Alternate names: CD174, FT3B, FucT-III, LE, Les, MGC131739ApplicationWB. Species reactivity: Cow, Dog, Human
Catalog No. 50-288-126