Learn More
Aviva Systems Biology Corp FUT6 Antibody - C-terminal region (ARP48467_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48467_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUT6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FUT6. Alternate names: FLJ40754, FT1A, FCT3A, Fuc-TVI, FucT-VIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53555