Learn More
Aviva Systems Biology Corp FUT8 Antibody - N-terminal region (ARP60784_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60784_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUT8. It was validated on Western Blot. . Alternate names: MGC26465ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-287-162