Learn More
Medchemexpress LLC Futibatinib | 1448169-71-8 | MFCD29037352 | 99.9% | 418.45 g/mol | C22H22N6O3 | 5 MG
SDP
Catalog No. 501871762
Encompass_Preferred
Futibatinib is an orally bioavailable, irreversible FGFR1-4 inhibitor supplied as a research-grade powder intended for analytical and preclinical laboratory research. It targets fibroblast growth factor receptors and shows low-nanomolar potency against FGFR1-4, including activity against several FGFR2 mutants.
  • Potent irreversible FGFR inhibitor with low-nanomolar activity.
  • Active against wild-type and mutant FGFR2 variants.
  • Supplied as a research-grade powder for analytical applications.
  • Molecular formula C22H22N6O3 and molecular weight 418.45 g/mol.
  • Intended for laboratory research use only; not for human or clinical use.
Catalog No. 50-187-1762 Supplier Medchemexpress LLC Supplier No. HY1008185MG
Futibatinib is an orally bioavailable, irreversible FGFR1-4 inhibitor supplied as a research-grade powder intended for analytical and preclinical laboratory research. It targets fibroblast growth factor receptors and shows low-nanomolar potency against FGFR1-4, including activity against several FGFR2 mutants.
  • Potent irreversible FGFR inhibitor with low-nanomolar activity.
  • Active against wild-type and mutant FGFR2 variants.
  • Supplied as a research-grade powder for analytical applications.
  • Molecular formula C22H22N6O3 and molecular weight 418.45 g/mol.
  • Intended for laboratory research use only; not for human or clinical use.