Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Futibatinib (TAS-120) | 1448169-71-8 | 418.45 g/mol | C22H22N6O3 | 100 MG
SDP

Catalog No. 5000212002
Encompass_Preferred

Futibatinib (TAS-120) is an orally bioavailable, irreversible fibroblast growth factor receptor (FGFR) inhibitor used in research to study FGFR-driven signaling and oncology models. It inhibits FGFR1-4 and retains activity against several FGFR2 mutants, making it a tool compound for in vitro and in vivo pathway inhibition studies.

  • Molecular weight: 418.45 g/mol.
  • CAS number: 1448169-71-8.
  • Chemical formula: C22H22N6O3.
  • Target: FGFR1-4, including mutant FGFR2 variants.
  • Potency: low-nanomolar IC50 values against FGFRs.
  • Typical applications: biochemical assays and cellular or animal studies of FGFR signaling.

Catalog No. 50-002-12002 Supplier Medchemexpress LLC Supplier No. HY100818100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Futibatinib (TAS-120) is an orally bioavailable, irreversible fibroblast growth factor receptor (FGFR) inhibitor used in research to study FGFR-driven signaling and oncology models. It inhibits FGFR1-4 and retains activity against several FGFR2 mutants, making it a tool compound for in vitro and in vivo pathway inhibition studies.

  • Molecular weight: 418.45 g/mol.
  • CAS number: 1448169-71-8.
  • Chemical formula: C22H22N6O3.
  • Target: FGFR1-4, including mutant FGFR2 variants.
  • Potency: low-nanomolar IC50 values against FGFRs.
  • Typical applications: biochemical assays and cellular or animal studies of FGFR signaling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.