Learn More
Selleck Chemical LLC Futibatinib2mg
SDP
Supplier:  Selleck Chemical LLC S88482MG
Encompass_Preferred
Futibatinib (TAS-120) is an irreversible fibroblast growth factor receptor (FGFR) inhibitor which inhibits all 4 subtypes of FGFR with enzyme IC50 values of 1.8 nM, 1.4 nM, 1.6 nM and 3.7 nM for FGFR1, FGFR2, FGFR3 and FGFR4, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3402