Learn More
Merit Medical Futura™ Safety Scalpel
Catalog No. 23306752
Change view
Click to view available options
Size:
# 10
# 11
# 15
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-306-752 # 15
23-306-750 # 10
23-306-751 # 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-306-752 Supplier Merit Medical Supplier No. SMS215
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to protect healthcare workers from accidental cuts associated with traditional scalpels
Retractable blades; color-coded.
  • Clinician controls the in-use or hidden position of the blade via a locking inset
  • Completely disposable, the scalpel's compact design conserves disposal container space
Order Info
Blades are precision sharpened and available in multiple sizes to accommodate various applications.
Specifications
Product Type Safety Scalpel
For Use With (Application) To Protect Healthcare Workers from Accidental Cuts associated with Traditional Scalpels
Size # 15
Disposable Single-use
Sterility Non-sterile
Handle Material Polystyrene