Learn More
Labrepco Inc Futura Silver Series PRIME 4.2 Cu. Ft. Undercounter Laboratory Freezer
SDP
Catalog No. NC1846169
Encompass_Preferred
The LHP-5-UFMBSS features one solid, stainless steel door with a recessed handle, stainless steel exterior, microprocessor temperature controller with audible and visual alarms, digital temperature display, remote alarm contacts and leveling legs.
Catalog No. NC1846169 Supplier Labrepco Inc Supplier No. LHP5UFMBSS
The LHP-5-UFMBSS features one solid, stainless steel door with a recessed handle, stainless steel exterior, microprocessor temperature controller with audible and visual alarms, digital temperature display, remote alarm contacts and leveling legs.