Learn More
Labrepco Inc Futura Silver Series PRIME 72 Cu. Ft. Laboratory Refrigerator, 3 Stainless Steel Doors, Microprocessor Temperature Control, 72 cu. ft.
SDP
Catalog No. NC3664325
Encompass_Preferred
The Futura Silver Series PRIME 72 Cu. Ft. Laboratory Refrigerator offers unparalleled capacity and precise temperature control, making it ideal for scientific environments. Constructed with durable stainless steel, it features a microprocessor temperature controller, self-closing doors with key locks, and robust security alarms, ensuring maximum sample safety. The unit's mobility is enhanced by field-installable swivel casters, setting it apart as a reliable choice for laboratory cooling needs.
Catalog No. NC3664325 Supplier Labrepco Inc Supplier No. LHP72SDSS
Add to Cart
Add to Cart
Request a Quote
The Futura Silver Series PRIME 72 Cu. Ft. Laboratory Refrigerator offers unparalleled capacity and precise temperature control, making it ideal for scientific environments. Constructed with durable stainless steel, it features a microprocessor temperature controller, self-closing doors with key locks, and robust security alarms, ensuring maximum sample safety. The unit's mobility is enhanced by field-installable swivel casters, setting it apart as a reliable choice for laboratory cooling needs.