Learn More
Labrepco Inc LabRepCo Futura Silver 21Cu Ft Freezer, Manual Defrost, Temperature Range -10°C to -25°C, 20 cu. ft.
SDP
Catalog No. NC2174295
Encompass_Preferred
The LabRepCo Futura Silver Series freezer is a reliable 20 cu. ft. upright laboratory/medical freezer designed for stable and efficient cold storage in clinical environments. With precise microprocessor controls and manual defrost, it ensures optimal sample integrity. Its durable construction, including galvanized steel interiors and adjustable wire shelves, sets it apart from competitors, making it ideal for demanding research applications.
This item is not returnable. View return policy
Catalog No. NC2174295 Supplier Labrepco Inc Supplier No. L2X20FM
The LabRepCo Futura Silver Series freezer is a reliable 20 cu. ft. upright laboratory/medical freezer designed for stable and efficient cold storage in clinical environments. With precise microprocessor controls and manual defrost, it ensures optimal sample integrity. Its durable construction, including galvanized steel interiors and adjustable wire shelves, sets it apart from competitors, making it ideal for demanding research applications.