Learn More
Anton Paar Future-proof ViscoQC with accessories for temperature control and diverse samples. Programmable methods and graphical analysis (flow curves, yield point). Fully compliant with 21 CFR Part 11, GMP, and GAMP 5 in one day.
Supplier:  Anton Paar 105024
Encompass_Preferred
Visco QC 300R Specifications:Intuitive user interface: 7" color touchscreen display; Options: keyboard, mouse, 2D bar code readerViscosity test: Multi Point ViscometerViscosity range [mPa·s or cP](depends on measuring system)R= 1.3 to 40 MSpeed [rpm]: 0.01 rpm to 250 rpmSpeed list with 18 standard speedsFreely selectable speeds within the speed rangeMax. spring torque [mNm]: 0.7187Accuracy: ±1.0 % full scale rangeRepeatability: ±0.2 % full scale rangeFeatures:Built-in Digital LevelMagnectic Coupling SpindlesAutomatic Measuring Range CalculationAutomatic Speed Searching (TruMode)Guided Spindle CorrectionFlow Curve (Optional)Yeld Point (Optionl)Gel Time (Optional)Accessories Available:Heli-PlusPTD 100 Temperature Control for Cone-PlateETD 300 Electrica Temperature Control for DIN SpindlesPTD 80 - Peltier Temperature Control for DIN SpindlesPTD 175 - Peltier Temperature Control for DIN Spindles
Catalog No. 50-005-80529