Learn More
Aviva Systems Biology Corp FUZ Antibody - C-terminal region (ARP65080_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65080_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUZ. It was validated on Western Blot. . Alternate names: FLJ22688, FYApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-288-424