Learn More
Novus Biologicals™ FUZ Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 80199 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-24750. For further blocking peptide related protocol, click here . |
| Gene Alias | FLJ22688, fuzzy homolog (Drosophila), FY, protein fuzzy homolog |
| Gene Symbol | FUZ |
| Quantity | 0.1 mg |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.