Learn More
Thomson Instrument Company eXtreme | FV Filter Vial, PVDF 0.2µm, Pre-Slit Septum, Red Cap
SDP
Supplier:  Thomson Instrument Company 85531200
Encompass_Preferred
Thomson eXtreme|FV Filter Vials put multi-layer depth filtration inside a standard autosampler vial, turning particulate-laden or viscous samples into clean, ready-to-inject filtrate in ~15 seconds. By separating compounds from the matrix, they deliver higher signal-to-noise and sharper peaks—while replacing syringe filters, and extra vials/caps.
•Multi-layer depth filtration — tackles viscous, particulate-rich samples (up to ~30% solids) with less fuss.
•Matrix clean-up in-vial — separates compounds from the matrix for higher S/N and better peak shape.
•Fast, simple workflow — raw sample to autosampler-ready in ~15 seconds.
•Replace SPE steps — swap labor-intensive SPE with a quick, lower-cost in-vial filtration.
•Fewer consumables — eliminates syringes, syringe filters, extra LC vials & caps; less plastic waste.
•Autosampler universal — standard LC-vial footprint, including UHPLC.
• Made in the USA
Catalog No. 50-396-706