Learn More
Innovative Research Inc FVIIDfcnt Hmn Pla 1ml
Catalog No. 502744592
Encompass_Preferred
Factor VII (FVII) Deficient Human Plasma 1ml - Factor VII (FVII) Deficient Human Plasma from Innovative Research is prepared from high-quality, citrated pooled human plasma that has been immuno-depleted of Factor VII so that the remaining activity is less than or equal to 1%.
Catalog No. 50-274-4592 Supplier Innovative Research Inc Supplier No. IFVIIDP1ML
Factor VII (FVII) Deficient Human Plasma 1ml - Factor VII (FVII) Deficient Human Plasma from Innovative Research is prepared from high-quality, citrated pooled human plasma that has been immuno-depleted of Factor VII so that the remaining activity is less than or equal to 1%.