Learn More
Medchemexpress LLC FX-06 1mg | 88650-17-3 | 1MG
SDP
Catalog No. 5000242895
Encompass_Preferred
FX-06 (Fibrin-derived peptide B 15-42) is a fibrin Bbeta chain-derived peptide FX-06 binds to VE-cadherin and inhibits leukocyte transmigration and initiates VE-cadherin-mediated signaling FX-06 can be used in the research of ischemia/reperfusion injury Dengue shock syndrome (DSS)[1 [2 [4
Catalog No. 50-002-42895 Supplier Medchemexpress LLC Supplier No. HY1062751MG
FX-06 (Fibrin-derived peptide B 15-42) is a fibrin Bbeta chain-derived peptide FX-06 binds to VE-cadherin and inhibits leukocyte transmigration and initiates VE-cadherin-mediated signaling FX-06 can be used in the research of ischemia/reperfusion injury Dengue shock syndrome (DSS)[1 [2 [4