Learn More
Medchemexpress LLC HY-16214 10mg , FX-11 CAS:213971-34-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1621410MG
Encompass_Preferred
HY-16214 10mg FX-11 CAS: 213971-34-7 FX-11 is a potent, selective, reversible and competitive lactate dehydrogenase A (LDHA) specific inhibitor, with a Ki of 8 μM. FX-11 can effectively activate PKM2 (pyruvate kinase M2). FX-11 reduces ATP levels and induces oxidative stress, ROS production and cell death. FX-11 shows antitumor activity in lymphoma and pancreatic cancer xenografts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9547