Learn More
Medchemexpress LLC HY-16214 5mg Medchemexpress, FX-11 CAS:213971-34-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY162145MG
Encompass_Preferred
Medchemexpress, HY-16214 5mg FX-11 CAS:213971-34-7 FX-11 is a potent lactate dehydrogenase A ( LDHA ) inhibitor with an IC 50 of 23.3 μM for HeLa cell,a K i value of 8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2763