Learn More
Selleck Chemical LLC FX1 5mg 1426138-42-2
SDP
Catalog No. 501948100
Encompass_Preferred
FX1 is a selective BCL6 BTB inhibitor with an IC50 value of 35 ?M in reporter assays. FX1 shows great selectivity against a panel of 50 different kinases. 10 ?M FX1 fails to significantly inhibit of any of these kinases. FX1 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8100 Supplier Selleck Chemical LLC Supplier No. S85915MG
FX1 is a selective BCL6 BTB inhibitor with an IC50 value of 35 ?M in reporter assays. FX1 shows great selectivity against a panel of 50 different kinases. 10 ?M FX1 fails to significantly inhibit of any of these kinases. FX1 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.