Learn More
Hoefer, Inc FXGD3 imaging Unit
SDP
Catalog No. NC3655198
Encompass_Preferred
"FXGD3 gel doc system with high resolution camera for all fluorescence requirements.Versatile high-resolution gel docAn entry-level system for imaging fluorescence and visible applications, the FXGD3 gel doc is the ideal solution for your laboratory. Using your choice of dyes or stains, the FXGD3 automatically selects the proper lighting and filters to detect close bands on both small and large gels. This system is the ultimate in gel doc flexibility when choosing white light options for Coomassie Blue, UV and blue lighting options, ethidium bromide or SYBR® Safe gels.The stylish and modular designed FXGD3 gel doc system includes a high resolution 5m pixel camera which is capable of giving outstanding images with incredible spatial resolution as well as a motor-driven zoom lens and a motor-driven filter wheel and feedback lens, allowing you to optimise your image capture using user protocols.
Catalog No. NC3655198 Supplier Hoefer, Inc Supplier No. FXGD3 UNIT
Add to Cart
Add to Cart
Request a Quote
"FXGD3 gel doc system with high resolution camera for all fluorescence requirements.Versatile high-resolution gel docAn entry-level system for imaging fluorescence and visible applications, the FXGD3 gel doc is the ideal solution for your laboratory. Using your choice of dyes or stains, the FXGD3 automatically selects the proper lighting and filters to detect close bands on both small and large gels. This system is the ultimate in gel doc flexibility when choosing white light options for Coomassie Blue, UV and blue lighting options, ethidium bromide or SYBR® Safe gels.The stylish and modular designed FXGD3 gel doc system includes a high resolution 5m pixel camera which is capable of giving outstanding images with incredible spatial resolution as well as a motor-driven zoom lens and a motor-driven filter wheel and feedback lens, allowing you to optimise your image capture using user protocols.